Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RGPD5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5104056
Description
Antibody detects endogenous levels of total RGPD5.
The RANBP2-like and GRIP domain containing 5 protein (RGPD5) has high similarity to RANBP2, a large RAN-binding protein localized at the cytoplasmic side of the nuclear pore complex. The gene coding for RGPD5 is thought to have arisen from a gene duplication event of RANBP2 as these highly homologous genes are located close to each other at chromosome 2q11-q12. RGPD5 was identified as an HIV dependency factor (HDF), suggesting that RGPD5 may be an important drug target in HIV treatment. At least two isoforms of RGPD5 are known to exist, of which the shorter isoform is expressed primarily in testis, while the longer of the two is expressed at low levels in a number of somatic tissues.
Specifications
RGPD5 | |
Polyclonal | |
Unconjugated | |
RGPD5 | |
BS63; BS-63; epididymis luminal protein 161; HEL161; ran-binding protein 2-like 1; ran-binding protein 2-like 1/2; RANBP2L1; RANBP2L2; ranBP2-like 1; ranBP2-like 1/2; RANBP2-like and GRIP domain containing 5; RANBP2-like and GRIP domain-containing protein 5; RANBP2-like and GRIP domain-containing protein 5/6; RANBPL1; RGP5; RGP6; RGP7; RGPD5; RGPD6; RGPD7; Sperm membrane protein BS-63 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
84220 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q99666 | |
RGPD5 | |
A synthesized peptide derived from human RGPD5; RGPD6(Accession Q99666), corresponding to amino acid residues N204-E254. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction