Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RGS10 Monoclonal Antibody (OTI1B10), TrueMAB™, OriGene
SDP

Catalog No. 502543711
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-3711 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-3711 Supplier OriGene Supplier No. CF812310
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RGS10 Monoclonal antibody specifically detects RGS10 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen RGS10
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1B10
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene TTC7A
Gene Accession No. Q8BGB2, Q9ULT0
Gene Alias 2310010N19Rik; regulator of G protein signaling 10; regulator of G-protein signaling 10; regulator of G-protein signalling 10; Rgs10; similar to Regulator of G-protein signaling 10 (RGS10)
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-450 of human TTC7A (NP_065191) produced in E.coli.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 225049, 362696, 57217
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.