Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RGS10 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA546819
Description
Peptide sequence: DQIFNLMKYD SYSRFLKSDL FLKHKRTEEE EEDLPDAQTA AKRASRIYNT Sequence homology: Cow: 81%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Rabbit: 90%; Rat: 90%.
RGS10 inhibits signal transduction by increasing the GTPASE activity of G protein alpha subunits thereby driving them into their inactive GDP-bound form. It associates specifically with the activated forms of the G protein subunits G(I)-ALPHA and G(Z)- alpha but fails to interact with the structurally and functionally distinct G(S)-alpha subunit. Activity on G(Z)-alpha is inhibited by palmitoylation of the G-protein.
Specifications
RGS10 | |
Polyclonal | |
Unconjugated | |
RGS10 | |
2310010N19Rik; regulator of G protein signaling 10; regulator of G-protein signaling 10; regulator of G-protein signalling 10; Rgs10; similar to Regulator of G-protein signaling 10 (RGS10) | |
Rabbit | |
Affinity Chromatography | |
RUO | |
6001, 67865 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O43665, Q9CQE5 | |
RGS10 | |
Synthetic peptide directed towards the middle region of human RGS10. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction