Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RGS10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA575559
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA575559 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA575559 Supplier Invitrogen™ Supplier No. PA575559
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

RGS10 inhibits signal transduction by increasing the GTPASE activity of G protein alpha subunits thereby driving them into their inactive GDP-bound form. It associates specifically with the activated forms of the G protein subunits G(I)-ALPHA and G(Z)- alpha but fails to interact with the structurally and functionally distinct G(S)-alpha subunit. Activity on G(Z)-alpha is inhibited by palmitoylation of the G-protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RGS10
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene RGS10
Gene Accession No. O43665
Gene Alias 2310010N19Rik; regulator of G protein signaling 10; regulator of G-protein signaling 10; regulator of G-protein signalling 10; Rgs10; similar to Regulator of G-protein signaling 10 (RGS10)
Gene Symbols RGS10
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 91-140 of Human RGS10.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6001
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.