Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RGS10 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569199
Description
This target displays homology in the following species: Cow: 93%; Human: 93%; Mouse: 100%; Pig: 93%; Rat: 100%.
RGS10 inhibits signal transduction by increasing the GTPASE activity of G protein alpha subunits thereby driving them into their inactive GDP-bound form. It associates specifically with the activated forms of the G protein subunits G(I)-ALPHA and G(Z)- alpha but fails to interact with the structurally and functionally distinct G(S)-alpha subunit. Activity on G(Z)-alpha is inhibited by palmitoylation of the G-protein.
Specifications
RGS10 | |
Polyclonal | |
Unconjugated | |
RGS10 | |
2310010N19Rik; regulator of G protein signaling 10; regulator of G-protein signaling 10; regulator of G-protein signalling 10; Rgs10; similar to Regulator of G-protein signaling 10 (RGS10) | |
Rabbit | |
Affinity chromatography | |
RUO | |
67865 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9CQE5 | |
RGS10 | |
synthetic peptide corresponding to a region of mouse RGS10 (aa 1-50). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction