Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RGS11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560210
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560210 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560210 Supplier Invitrogen™ Supplier No. PA560210
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GDRLVIACQE QTYWLVNRPP PGAPDVLEQG PGRGSCAASR VLMTKSADFH KREIEYFRKA LGRTRVKSSV CLEAYLSFCG QRGPHDPL Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 38%.

The protein encoded by this gene belongs to the RGS (regulator of G protein signaling) family. Members of the RGS family act as GTPase-activating proteins on the alpha subunits of heterotrimeric, signal-transducing G proteins. This protein inhibits signal transduction by increasing the GTPase activity of G protein alpha subunits, thereby driving them into their inactive GDP-bound form. Alternative splicing occurs at this locus and two transcript variants encoding distinct isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RGS11
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RGS11
Gene Accession No. O94810
Gene Alias AC004754.2; C78048; regulator of G-protein signaling 11; regulator of G-protein signalling 11; RGS11; RS11
Gene Symbols RGS11
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RGS11. Recombinant protein control fragment (Product #RP-98614).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8786
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.