Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RGS13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110290
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110290 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110290 Supplier Invitrogen™ Supplier No. PA5110290
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSRRNCWICK MCRDESKRPP SNLTLEEVLQ WAQSFENLMA TKYGPVVYAA YLKMEHSDEN IQFWMACETY KKIASRWSRI SRAKKLYKIY IQPQSPREIN IDSSTRETII RNIQEPTETC FEEAQKIVYM HMERDSYPRF LKSEMYQKLL KTMQSNNSF.

RGS13 is the smallest member of the mammalian gene family, RGS (regulators of G protein-signaling) which are structural and functional homologues of yeast SST2 and C. elegans egl-10 proteins. RGS proteins attenuate G protein-mediated pathways by acting as GTPase-activating proteins (GAPs) for Ga subunits. RGS13 has been found most abundantly in human tonsil, thymus, lung, lymph node, and spleen. RGS13 regulate Gai, Gaq, and Gas coupled signaling cascades. It is a GTPase-activating protein for Gai and Gao. The protein blocks MAPK activity induced by Gai- or Gaq-coupled receptors. The protein also attenuates GTPase-deficient Gaq (GaqQL) mediated cAMP response element activation. The gene encoding for human RGS13 is localized on chromosome 1q31.2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RGS13
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.65 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RGS13
Gene Accession No. Q8K443
Gene Alias MGC17173; regulator of G protein signaling 13; regulator of G-protein signaling 13; regulator of G-protein signalling 13; RGD1562103; Rgs13; RP11-92K2.1
Gene Symbols RGS13
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-159 of human RGS13 (NP_002918.1).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 246709, 498246
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.