Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RGS14 Mouse anti-Mouse, Rat, Clone: N133/35, MilliporeSigma™

Catalog No. MABN185525 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN185525 25 μL
MABN1855100 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABN185525 Supplier MilliporeSigma Supplier No. MABN185525UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RGS14 Monoclonal antibody specifically detects RGS14 in Mouse, Rat samples. It is validated for Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RGS14
Applications Western Blot
Classification Monoclonal
Clone N133/35
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Host Species Mouse
Immunogen Full-length recombinant rat Regulator of G-protein signaling 14 (RGS14).
Purification Method Protein G purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse, Rat
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.