Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RGS16 Antibody (OTI2C9), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273884F
Description
RGS16 Monoclonal specifically detects RGS16 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
RGS16 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
RGS16 | |
Full length recombinant protein of human RGS16 (NP_002919) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Immunocytochemistry, Immunofluorescence | |
OTI2C9 | |
Immunocytochemistry/Immunofluorescence | |
A28-RGS14, A28-RGS14P, hRGS-r, regulator of G-protein signaling 16, regulator of G-protein signalling 16, Retinally abundant regulator of G-protein signaling, Retinal-specific RGS, RGSR, RGS-R | |
Mouse | |
Affinity Purified | |
Signal Transduction | |
6004.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction