Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RGS16 Monoclonal Antibody (OTI4E5)
GREENER_CHOICE

Catalog No. PIMA525803
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525803 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525803 Supplier Invitrogen™ Supplier No. MA525803
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RGS16 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow

The protein encoded by this gene belongs to the 'regulator of G protein signaling' family. It inhibits signal transduction by increasing the GTPase activity of G protein alpha subunits. It also may play a role in regulating the kinetics of signaling in the phototransduction cascade.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RGS16
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI4E5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RGS16
Gene Accession No. O15492, P97428
Gene Alias A28-RGS14; A28-RGS14P; hRGS-r; regulator of G-protein signaling 16; regulator of G-protein signalling 16; Retinally abundant regulator of G-protein signaling; retinal-specific RGS; Rgs14; RGS16; RGSR; RGS-R
Gene Symbols RGS16
Host Species Mouse
Immunogen Full length human recombinant protein of RGS16 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19734, 6004
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.