Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RGS17 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP133694
Description
RGS17 Polyclonal specifically detects RGS17 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
RGS17 | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
RGS17 | |
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human RGS17. The exact sequence is proprietary. | |
100 μL | |
Apoptosis, Signal Transduction | |
26575 | |
Human | |
IgG |
Western Blot, Immunocytochemistry, Immunofluorescence | |
1.0mg/mL | |
Western Blot 1:1000-1:10000, Immunocytochemistry/Immunofluorescence 1:100-1:1000 | |
hRGS17, regulator of G-protein signaling 17, regulator of G-protein signalling 17, RGS-17, RGSZ2 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Tested in human. Not yet tested in any other species. | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction