Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RGS17 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584296
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584296 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584296 Supplier Invitrogen™ Supplier No. PA584296
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RDSFPRFLNS QIYKSFVEST AGSSSES Highest antigen sequence indentity to the following orthologs: Mouse - 81%, Rat - 89%.

This gene encodes a member of the regulator of G-protein signaling family. This protein contains a conserved, 120 amino acid motif called the RGS domain and a cysteine-rich region. The protein attenuates the signaling activity of G-proteins by binding to activated, GTP-bound G alpha subunits and acting as a GTPase activating protein (GAP), increasing the rate of conversion of the GTP to GDP. This hydrolysis allows the G alpha subunits to bind G beta/gamma subunit heterodimers, forming inactive G-protein heterotrimers, thereby terminating the signal.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RGS17
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RGS17
Gene Accession No. Q9UGC6
Gene Alias 6430507P11Rik; hRGS17; regulator of G-protein signaling 17; regulator of G-protein signaling Z2; regulator of Gz-selective protein signaling 2; Rgs17; RGS-17; Rgsz2
Gene Symbols RGS17
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RGS17. Recombinant protein control fragment (Product #RP-105046).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26575
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.