Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RGS19 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578315
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578315 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578315 Supplier Invitrogen™ Supplier No. PA578315
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: human RGS19-transfected 293T Predicted Reactivity: Mouse (87%), Rat (87%), Rhesus Monkey (96%), Bovine (84%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

RGS19 inhibits signal transduction by increasing the GTPase activity of G protein alpha subunits thereby driving them into their inactive GDP-bound form. Binds to G-alpha subfamily 1 members, predominantly to G(i)-alpha-3. Activity on G(z)-alpha is inhibited by phosphorylation and palmitoylation of the G-protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RGS19
Applications Western Blot
Classification Polyclonal
Concentration 0.28 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene RGS19
Gene Accession No. P49795
Gene Alias 2610042F04Rik; AI324841; AW547781; calcium/calmodulin-dependent protein kinase I; Camki; G alpha interacting protein; G protein signalling regulator 19; GAIP; GAIP protein; G-alpha interacting protein; G-alpha-interacting protein; GNAI3IP; GS19; guanine nucleotide binding protein alpha inhibiting activity polypeptide 3 interacting protein; regulator of G-protein signaling 19; regulator of G-protein signalling 19; Rgs19; RGSGAIP
Gene Symbols RGS19
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human RGS19. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10287
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.