Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RGS2 Polyclonal Antibody

RGS2 Polyclonal Antibody
Supplier: Thermo Scientific PA519644
Description
RGS2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Specifications
| RGS2 | |
| Polyclonal | |
| Unconjugated | |
| RGS2 | |
| cell growth-inhibiting gene 31 protein; cell growth-inhibiting protein 31; G0 to G1 switch regulatory 8, 24kD; G0/G1 switch regulatory protein 8; G0S8; GIG31; GOS8; regulator of G protein signaling 2; regulator of G protein signaling 2; regulator of G-protein signaling 2; regulator of G-protein signaling 2; regulator of G-protein signaling 2, 24kDa; regulator of G-protein signaling protein 2; regulator of G-protein signalling 2, 24kDa; Rgs2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 488584, 5997 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.6 mg/mL | |
| PBS with 1% BSA and 0.02% sodium azide; pH 7.4 | |
| P41220 | |
| RGS2 | |
| Synthetic peptide conjugated to KLH derived from within residues 150 to the C-terminus of Human RGS2. | |
| 100 μg | |
| Primary | |
| Human, Canine | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction