Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RGS3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA126488
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA126488 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA126488 Supplier Invitrogen™ Supplier No. PA126488
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-26488 detects RGS3 from human samples.

This gene encodes a member of the regulator of G-protein signaling (RGS) family. This protein is a GTP-ase activating protein which inhibits G-protein mediated signal transduction. The protein is largely cytosolic, but G-protein activation leads to translocation of this protein to the plasma membrane. A nuclear form of this protein has also been described, but its sequence has not been identified. Multiple alternatively spliced transcript variants have been described for this gene but the full-length nature of some transcripts is not yet known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RGS3
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene RGS3
Gene Accession No. P49796
Gene Alias 4930506N09Rik; C2 membrane binding, PDZ protein/protein interaction, ATP/GTP binding domains; C2pa; C2PA-RGS3; FLJ20370; FLJ31516; FLJ90496; PDZ-RGS3; regulator of G-protein signaling 3; regulator of G-protein signalling 3; RGP3; RGS3; RGS3S; SRB-RGS
Gene Symbols RGS3
Host Species Rabbit
Immunogen Recombinant fragment-amino acids 401-519 of human RGS 3.
Purification Method Protein A
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5998
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.