Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RGS4 Recombinant Rabbit Monoclonal Antibody (HL1522)
GREENER_CHOICE

Catalog No. PIMA547078
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547078 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547078 Supplier Invitrogen™ Supplier No. MA547078
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial.

Regulator of G protein signaling (RGS) family members are regulatory molecules that act as GTPase activating proteins (GAPs) for G alpha subunits of heterotrimeric G proteins. RGS proteins are able to deactivate G protein subunits of the Gi alpha, Go alpha and Gq alpha subtypes. They drive G proteins into their inactive GDP-bound forms. Regulator of G protein signaling 4 belongs to this family. All RGS proteins share a conserved 120-amino acid sequence termed the RGS domain. Regulator of G protein signaling 4 protein is 37% identical to RGS1 and 97% identical to rat Rgs4. This protein negatively regulate signaling upstream or at the level of the heterotrimeric G protein and is localized in the cytoplasm. Alternatively spliced transcript variants have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RGS4
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone HL1522
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene RGS4
Gene Accession No. O08899, P49798, P49799
Gene Alias AA004315; AA597169; ESTM48; ESTM50; MGC2124; MGC60244; regulator of G protein signaling 4; regulator of G protein signaling 4; regulator of G-protein signaling 4; regulator of G-protein signaling 4; regulator of G-protein signalling 4; RGP4; RGS4; RP11-430G6.1; schizophrenia disorder 9; SCZD9; similar to Regulator of G-protein signaling 4 (RGS4) (RGP4)
Gene Symbols RGS4
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human RGS4. The exact sequence is proprietary.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19736, 29480, 5999
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.