Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RGS4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104096
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104096 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104096 Supplier Thermo Scientific Supplier No. PA5104096
Add to Cart
Add to Cart

RGS4 Polyclonal Antibody

RGS4 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen RGS4
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene RGS4
Gene Accession No. O08899, P49798, P49799
Gene Alias AA004315; AA597169; ESTM48; ESTM50; MGC2124; MGC60244; regulator of G protein signaling 4; regulator of G protein signaling 4; regulator of G-protein signaling 4; regulator of G-protein signaling 4; regulator of G-protein signalling 4; RGP4; RGS4; RP11-430G6.1; schizophrenia disorder 9; SCZD9; similar to Regulator of G-protein signaling 4 (RGS4) (RGP4)
Gene Symbols RGS4
Host Species Rabbit
Immunogen A synthesized peptide derived from human RGS4(Accession P49798), corresponding to amino acid residues K162-A205.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19736, 29480, 5999
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.