Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RGS5 Antibody (OTI1C1), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273887AF350
Description
RGS5 Monoclonal specifically detects RGS5 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.Specifications
RGS5 | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
RGS5 | |
Full length recombinant protein of human RGS5 (NP_003608) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Flow Cytometry, Immunohistochemistry | |
OTI1C1 | |
Western Blot, Flow Cytometry, Immunohistochemistry | |
MST092, MST106, MST129, MSTP032, MSTP092, MSTP106, MSTP129, regulator of G-protein signaling 5, regulator of G-protein signalling 5 | |
Mouse | |
Affinity Purified | |
Signal Transduction | |
8490.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction