Learn More
Invitrogen™ RGS5 Monoclonal Antibody (OTI1C1)

Description
RGS5 Monoclonal Antibody for Flow, ICC/IF, IHC, IHC (P), Western Blot
Specifications
Specifications
| Antigen | RGS5 |
| Applications | Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | OTI1C1 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, 1% BSA and 0.02% sodium azide; pH 7.3 |
| Gene | RGS5 |
| Gene Accession No. | O15539 |
| Gene Alias | 1110070A02Rik; MST092; MST106; MST129; MSTP032; MSTP092; MSTP106; MSTP129; regulator of G protein signaling 5; regulator of G-protein signaling 5; regulator of G-protein signalling 5; RGS5; RGS-5; RP11-430G6.2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.