Learn More
RGS5 Mouse anti-Human, Clone: OTI1C1, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF503075

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The regulator of G protein signaling (RGS) proteins are signal transduction molecules that have structural homology to SST2 of Saccharomyces cerevisiae and EGL-10 of Caenorhabditis elegans. Multiple genes homologous to SST2 are present in higher eukaryotes. RGS proteins are involved in the regulation of heterotrimeric G proteins by acting as GTPase activators. The mRNA of the human gene is abundantly expressed in heart, lung, skeletal muscle, and small intestine, and at low levels in brain, placenta, liver, colon, and leukocytes.Specifications
RGS5 | |
Monoclonal | |
Unconjugated | |
RGS5 | |
MST092, MST106, MST129, MSTP032, MSTP092, MSTP106, MSTP129 | |
Mouse | |
Affinity Chromatography | |
RUO | |
8490 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
OTI1C1 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
O15539 | |
RGS5 | |
Full length human recombit protein of human RGS5 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.