Learn More
RGS5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 115901AP150UL

Description
The 42 kDa band recognized by this antibody is a RGS5-RGS5 dimer.
The regulator of G protein signaling (RGS) proteins are signal transduction molecules that have structural homology to SST2 of Saccharomyces cerevisiae and EGL-10 of Caenorhabditis elegans. Multiple genes homologous to SST2 are present in higher eukaryotes. RGS proteins are involved in the regulation of heterotrimeric G proteins by acting as GTPase activators. The mRNA of the human gene is abundantly expressed in heart, lung, skeletal muscle, and small intestine, and at low levels in brain, placenta, liver, colon, and leukocytes.Specifications
RGS5 | |
Polyclonal | |
Unconjugated | |
RGS5 | |
MST092, MST106, MST129, MSTP032, MSTP092, MSTP106, MSTP129, RGS5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
19737, 54294, 8490 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O08850, O15539, P49800 | |
Rgs5 | |
RGS5 Fusion Protein Ag2196 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.