Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RGS6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530876
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530876 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530876 Supplier Invitrogen™ Supplier No. PA530876
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: PC-3. Predicted reactivity: Mouse (96%), Rat (97%), Xenopus laevis (81%), Chicken (92%), Rhesus Monkey (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Members of the RGS (regulator of G protein signaling) family, such as RGS6, modulate G protein function by activating the intrinsic GTPase activity of the alpha (guanine nucleotide-binding) subunits (Seki et al., 1999 [PubMed 10083744]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen RGS6
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene RGS6
Gene Accession No. P49758, P49801
Gene Alias DKFZp313G1241; FLJ43552; GAP; H_DJ1108A12.1; MGC142132; regulator of G-protein signaling 6; regulator of G-protein signalling 6; RGS6; S914; WUGSC:H_DJ0283M22.1; WUGSC:H_DJ1108A12.1
Gene Symbols RGS6
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human RGS6. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54295, 9628
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.