Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RGS7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA578150
Description
Positive Control: U87-MG, SK-N-SH Predicted Reactivity: Mouse (98%), Rat (98%), Chicken (95%), Bovine (98%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
RGS7, regulator of G protein signaling 6, belongs to a subfamily of RGS proteins, which serve as GAPs (GTPase-activating proteins), increasing the rate at which GTP is converted to GDP by isoform alpha G proteins.
Specifications
RGS7 | |
Polyclonal | |
Unconjugated | |
RGS7 | |
regulator of G protein signaling 7; regulator of G-protein signaling 7; Regulator of G-protein signaling 7 (RGS7); regulator of G-protein signaling RGS7; regulator of G-protein signalling 7; RGS7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6000 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
P49802 | |
RGS7 | |
Recombinant protein encompassing a sequence within the center region of human RGS7. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction