Learn More
Invitrogen™ RGS8 Polyclonal Antibody

Description
PA1-4262 detects the human RGS8 protein in human samples. PA1-4262 has been successfully used in Western blot procedures. PA1-4262 immunizing peptide corresponds to amino acid residues 1-16 of the human RGS8 protein.
Specifications
Specifications
| Antigen | RGS8 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | RGS8 |
| Gene Accession No. | P57771 |
| Gene Alias | 6530413N01Rik; Regulator of G-protein signaling 8; regulator of G-protein signalling 8; Rgs8 |
| Gene Symbols | RGS8 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.