Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RGS8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585008
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585008 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585008 Supplier Invitrogen™ Supplier No. PA585008
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PRFLRSKMYL DLLSQSQRRL Highest antigen sequence indentity to the following orthologs: Rat - 100%, Mouse - 100%.

A novel human RGS8 gene (homologue of the rat RGS8 gene) is expressed in prostate tissues and represents a positional candidate for the HPC1 locus, which is involved in hereditary prostate cancer and hyperparathyroidism-jaw tumor syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RGS8
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RGS8
Gene Accession No. P57771
Gene Alias 6530413N01Rik; Regulator of G-protein signaling 8; regulator of G-protein signalling 8; Rgs8
Gene Symbols RGS8
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RGS8. Recombinant protein control fragment (Product #RP-108670).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85397
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.