Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RGS8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597322
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597322 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597322 Supplier Invitrogen™ Supplier No. PA597322
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAALLMPRRN KGMRTRLGCL SHKSDSCSDF TAILPDKPNR ALKRLSTEEA TRWADSFDVL LSHKYGVAAF RAFLKTEFSE ENLEFWLACE EFKKTRSTAK LVSKAHRIFE EFVDVQAPRE VNIDFQTREA TRKNLQEPSL TCFDQAQGKV HSLMEKDSYP RFLRSKMYLD LLSQSQRRLS; Positive Samples: SH-SY5Y, Jurkat, Mouse brain, Rat brain.

A novel human RGS8 gene (homologue of the rat RGS8 gene) is expressed in prostate tissues and represents a positional candidate for the HPC1 locus, which is involved in hereditary prostate cancer and hyperparathyroidism-jaw tumor syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RGS8
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.93 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RGS8
Gene Accession No. P49804, P57771, Q8BXT1
Gene Alias 6530413N01Rik; Regulator of G-protein signaling 8; regulator of G-protein signalling 8; Rgs8
Gene Symbols RGS8
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-180 of human RGS8 (NP_0010959201).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54297, 67792, 85397
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.