Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RGS9 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP2932310.02ML
Description
RGS9 Polyclonal antibody specifically detects RGS9 in Mouse samples. It is validated for Western BlotSpecifications
| RGS9 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| MGC26458, PERRSMGC111763, regulator of G-protein signaling 9, regulator of G-protein signalling 9, RGS9L | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 230-350 of human RGS9 (NP_003826.2). MYYQQALMRSTVKSSVSLGGIVKYSEQFSSNDAIMSGCLPSNPWITDDTQFWDLNAKLVEIPTKMRVERWAFNFSELIRDPKGRQSFQYFLKKEFSGENLGFWEACEDLKYGDQSKVKEKA | |
| 0.02 mL | |
| Signal Transduction | |
| 8787 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction