Learn More
Description
MA1-723 detects RGS9-1 protein from bovine and mouse samples. MA1-723 has been successfully used in Western blot, immunocytochemistry and immunoprecipitation procedures. By Western blot, this antibody detects a ~57 kDa protein representing RGS-1 protein from bovine retinal extracts. The MA1-723 immunogen is full length, bacterially expressed RGS9-1 protein.
Specifications
Specifications
| Antigen | RGS9 |
| Applications | Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | D7 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1 mg/mL BSA and 0.05% sodium azide |
| Gene | RGS9 |
| Gene Accession No. | O46469, O54828 |
| Gene Alias | GTPase accelerating protein for heterotrimeric G-proteins; MGC111763; MGC26458; PERRS; regulator of G protein signaling 9; regulator of G protein signaling 9; regulator of G-protein signaling 9; regulator of G-protein signaling 9; regulator of G-protein signalling 9; Rgs9; RGS9-1; Rgs9-2; RGS9L; ROS GAP |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
