Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RGS9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546820
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546820 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546820 Supplier Invitrogen™ Supplier No. PA546820
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MYYQQALMRS TVKSSVSLGG IVKYSEQFSS NDAIMSGCLP SNPWITDDTQ Sequence homology: Cow: 100%; Dog: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

The cascade of biochemical reactions within the vision system is complex and contains numerous steps taking place in nanoseconds. RGS is a family of GTPase accelerating proteins (GAPs). RGS9 is a member of this family and is the predominant source of GAP activity in rod outer segments. RGS9-1, found predominately in vertebrate cone and rod photoreceptors, is a useful model for understanding how rapid recovery kinetics of the light responses in retinal photoreceptors take place.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RGS9
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RGS9
Gene Accession No. O75916
Gene Alias GTPase accelerating protein for heterotrimeric G-proteins; MGC111763; MGC26458; PERRS; regulator of G protein signaling 9; regulator of G protein signaling 9; regulator of G-protein signaling 9; regulator of G-protein signaling 9; regulator of G-protein signalling 9; Rgs9; RGS9-1; Rgs9-2; RGS9L; ROS GAP
Gene Symbols RGS9
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human RGS9 (aa 230-279).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8787
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.