Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RGS9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582497
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582497 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582497 Supplier Invitrogen™ Supplier No. PA582497
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LDAAQTHIYM LMKKDSYARY LKSPIYKDML AKAIEPQETT KKSSTLPFMR RHLRSSPSPV ILRQLEEEAK AREAANTVDI TQPGQHMAPS PHLTVYTGTC MPPSPSSPFS SSCRSPRKPF ASPSRFIRRP STTIC Highest antigen sequence indentity to the following orthologs: Mouse - 93%, Rat - 92%.

The cascade of biochemical reactions within the vision system is complex and contains numerous steps taking place in nanoseconds. RGS is a family of GTPase accelerating proteins (GAPs). RGS9 is a member of this family and is the predominant source of GAP activity in rod outer segments. RGS9-1, found predominately in vertebrate cone and rod photoreceptors, is a useful model for understanding how rapid recovery kinetics of the light responses in retinal photoreceptors take place.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RGS9
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RGS9
Gene Accession No. O75916
Gene Alias GTPase accelerating protein for heterotrimeric G-proteins; MGC111763; MGC26458; PERRS; regulator of G protein signaling 9; regulator of G protein signaling 9; regulator of G-protein signaling 9; regulator of G-protein signaling 9; regulator of G-protein signalling 9; Rgs9; RGS9-1; Rgs9-2; RGS9L; ROS GAP
Gene Symbols RGS9
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RGS9. Recombinant protein control fragment (Product #RP-105727).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8787
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.