Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RH5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144539
Description
RH5 Polyclonal Antibody for Western Blot, ELISA
ATP-dependent RNA helicase required for 60S ribosomal subunit synthesis. Involved in efficient pre-rRNA processing, predominantly at site A3, which is necessary for the normal formation of 25S and 5.8S rRNAs (By similarity).
Specifications
RH5 | |
Polyclonal | |
Unconjugated | |
RH5 | |
DEAD-box ATP-dependent RNA helicase | |
Rabbit | |
Affinity Chromatography | |
RUO | |
840087 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot | |
5 mg/mL | |
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
Q9C551 | |
RH5 | |
Recombinant Arabidopsis thaliana RH5 protein (53-142aa). | |
100 μL | |
Primary | |
Plant | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction