Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RHAG Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551097
Description
RHAG Polyclonal Antibody detects endogenous levels of RHAG protein.
Associated with rhesus blood group antigen expression. May be part of an oligomeric complex which is likely to have a transport or channel function in the erythrocyte membrane. {ECO:0000269; PubMed:11062476, ECO:0000269; PubMed:11861637}.
Specifications
RHAG | |
Polyclonal | |
Unconjugated | |
RHAG | |
Ammonium transporter Rh type A; CD241; Erythrocyte membrane glycoprotein Rh50; erythrocyte plasma membrane 50 kDa glycoprotein; OHS; OHST; Rh 50 glycoprotein; rh family type A glycoprotein; rh type A glycoprotein; RH2; RH50; Rh50A; Rh50GP; RHAG; Rh-associated glycoprotein; Rhesus associated polypeptide, 50-KD; rhesus blood group family type A glycoprotein; Rhesus blood group-associated ammonia channel; Rhesus blood group-associated glycoprotein; SLC42A1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6005 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1 mg/mL | |
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q02094 | |
RHAG | |
Synthesized peptide derived from the N-terminal region of human CD241. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction