Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RHAG Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563110
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563110 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563110 Supplier Invitrogen™ Supplier No. PA563110
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ETDQTVLEQL NITKPTDMGI FFELYP Highest antigen sequence identity to the following orthologs: Mouse - 35%, Rat - 37%.

Associated with rhesus blood group antigen expression. May be part of an oligomeric complex which is likely to have a transport or channel function in the erythrocyte membrane. {ECO:0000269; PubMed:11062476, ECO:0000269; PubMed:11861637}.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RHAG
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RHAG
Gene Accession No. Q02094
Gene Alias Ammonium transporter Rh type A; CD241; Erythrocyte membrane glycoprotein Rh50; erythrocyte plasma membrane 50 kDa glycoprotein; OHS; OHST; Rh 50 glycoprotein; rh family type A glycoprotein; rh type A glycoprotein; RH2; RH50; Rh50A; Rh50GP; RHAG; Rh-associated glycoprotein; Rhesus associated polypeptide, 50-KD; rhesus blood group family type A glycoprotein; Rhesus blood group-associated ammonia channel; Rhesus blood group-associated glycoprotein; SLC42A1
Gene Symbols RHAG
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RHAG. Recombinant protein control fragment (Product #RP-101735).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6005
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.