Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RHAMM Recombinant Rabbit Monoclonal Antibody (3L10H9)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA535150
Description
Immunogen sequence: MSFPKAPLKR FNDPSGCAPS PGAYDVKTLE VLKGPVSFQK SQRFKQQKES KQNLNVDKDT TLPASARKVK SSESKESQKN DKDLKILEKE IRVLLQERGA.
The protein encoded by this gene is involved in cell motility. It is expressed in breast tissue and together with other proteins, it forms a complex with BRCA1 and BRCA2, thus is potentially associated with higher risk of breast cancer. Alternatively spliced transcript variants encoding different isoforms have been noted for this gene.
Specifications
RHAMM | |
Recombinant Monoclonal | |
1.95 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O75330, Q00547 | |
HMMR | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human CD168/RHAMM (O75330). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
3L10H9 | |
Unconjugated | |
HMMR | |
AA386826; CD168; Hmmr; Hyaluronan mediated motility receptor; hyaluronan mediated motility receptor (RHAMM); hyaluronan-mediated motility receptor; hyaluronan-mediated motility receptor (RHAMM); IHABP; Intracellular hyaluronic acid-binding protein; receptor for hyaluronan-mediated motility; RHAMM | |
Rabbit | |
Affinity chromatography | |
RUO | |
15366, 3161 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction