Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RHAMM Polyclonal Antibody

RHAMM Polyclonal Antibody
Supplier: Thermo Scientific PA5111531
Description
RHAMM Polyclonal Antibody for ICC/IF
Specifications
| RHAMM | |
| Polyclonal | |
| Unconjugated | |
| HMMR | |
| AA386826; CD168; Hmmr; Hyaluronan mediated motility receptor; hyaluronan mediated motility receptor (RHAMM); hyaluronan-mediated motility receptor; hyaluronan-mediated motility receptor (RHAMM); IHABP; Intracellular hyaluronic acid-binding protein; receptor for hyaluronan-mediated motility; RHAMM | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 3161 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.60 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O75330 | |
| HMMR | |
| Recombinant Protein corresonding to Human RHAMM. Recombinant protein control fragment (Product #RP-107075). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction