Learn More
BD Rhapsody™ Package
Catalog No. bdb633707
Click to view available options
:
BD Rhapsody
This item is not returnable. View return policy
This item is not returnable. View return policy
This instrument allows for single-cell mRNA capture upstream of library preparation.
  • Allows for single-cell mRNA capture upstream of library preparation
  • Two custom pipettes are included that are required to run the instrument
  • Does not include reagents required to run the system
  • Does not include the BD Rhapsody™ Scanner
TRUSTED_SUSTAINABILITY
Specifications
Product Line BD Rhapsody
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.