Learn More
BD Rhapsody™ TCR/BCR Amplification Kit
Catalog No. BDB665345 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
BDB665345 1 Ea.
1 options
This item is not returnable. View return policy
Catalog No. BDB665345 Supplier BD Supplier No. 665345
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product
This item is not returnable. View return policy
The purpose of this kit is to provide the necessary reagents to prepare full length Human TCR and BCR libraries and is compatible with both Whole Transcriptome Analysis (WTA) and Targeted mRNA assays.
  • Provides the necessary reagents to prepare full length Human TCR and BCR libraries and is compatible with both Whole Transcriptome Analysis (WTA) and Targeted mRNA assays
  • In addition to this kit, an mRNA library preparation kit will be required, either the WTA amplification kit (cat no. 633801) or the Targeted mRNA & AbSeq Amp kit (cat no. 633774) with a targeted primer panel like the Human Immune Response panel (cat no. 633750). Cell capture (cat no. 664887 and 633733) and cDNA (cat no. 633773) reagents will also be needed
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Ea.