Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RHBDD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553102
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553102 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553102 Supplier Invitrogen™ Supplier No. PA553102
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PLKKIMEACA GGFSSSVGYP GRQYYFNSSG SSGYQDYYPH GRPDHYEEAP RNYDTYTAGL SEEEQLERAL QASLWDRGNT RNSPPPYGFH LSPEEMRRQR LHRFD Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 73%.

The Rhomboid family of proteins is made up of several widely conserved polytopic membrane serine proteases that play roles in growth and development. RHBDD1 is highly expressed in the testis and is involved in the cleavage of BIK, a proapoptotic member of the Bcl-2 family. Overexpression or suppression by RNAi of RHBDD1 in 293 cells will reduce or enhance BIK-mediated apoptosis, respectively, demonstrating that RHBDD1 modulates BIK-mediated apoptotic activity. In GC-1 cells, a spermatogonia cell line that can differentiate into spermatids within the seminiferous tubules, suppression of RHBDD1 expression by RNAi caused the cells to lose the ability to survive and differentiate in mouse seminiferous tubules, suggesting that RHBDD1 may be associated with mammalian spermatogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RHBDD1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RHBDD1
Gene Accession No. Q8TEB9
Gene Alias 4930418P06Rik; HSD50; HSD-50; mRHBDD1; MSD50; MSD-50; RGD1306477; RHBDD1; Rhbdl4; rhomboid domain containing 1; rhomboid domain-containing protein 1; Rhomboid-like protein 4; rhomboid-related protein 4; rRHBDD1; RRP4
Gene Symbols RHBDD1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RHBDD1. Recombinant protein control fragment (Product #RP-90982).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84236
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.