Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RHBDD2 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA562521
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562521 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562521 Supplier Thermo Scientific Supplier No. PA562521

Rabbit Polyclonal Antibody

Immunogen sequence: ERVALKLDQT FPFSLMRRIS VFKYVSGSSA ERRAAQSRKL NPVPGSYPTQ SCHPHLSPSH PVSQTQHASG QKLA Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 84%.

The Rhomboid family of proteins is made up of several widely conserved polytopic membrane serine proteases that play roles in growth and development. Little is known about the role of RHBDD2. Its overexpression in breast carcinoma appears to be an indicator of poor prognosis and may play a role in breast cancer progression. The related proteins RHBDD1 and RHBDD3 are both involved in the regulation of apoptosis, suggesting that RHBDD2 may also be involved in the modulation of apoptotic activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RHBDD2
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RHBDD2
Gene Accession No. Q6NTF9
Gene Alias 0610011L13Rik; AI480532; H_RG122E10.2a; H_RG122E10.2b; NPD007; Rhbdd2; Rhbdl7; rhomboid domain containing 2; rhomboid domain-containing protein 2; rhomboid, veinlet-like 7; rhomboid-like protein 7; upregulated during skeletal muscle growth 1; Usmg1; WUGSC:H_RG122E10.2a; WUGSC:H_RG122E10.2b
Gene Symbols RHBDD2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RHBDD2
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57414
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.