Learn More
Invitrogen™ RHBDD2 Polyclonal Antibody

Description
A suggested positive control is rat lung tissue lysate. PA5-20861 can be used with blocking peptide PEP-0975.
Specifications
Specifications
| Antigen | RHBDD2 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | RHBDD2 |
| Gene Accession No. | Q6NTF9, Q8VEK2 |
| Gene Alias | 0610011L13Rik; AI480532; H_RG122E10.2a; H_RG122E10.2b; NPD007; Rhbdd2; Rhbdl7; rhomboid domain containing 2; rhomboid domain-containing protein 2; rhomboid, veinlet-like 7; rhomboid-like protein 7; upregulated during skeletal muscle growth 1; Usmg1; WUGSC:H_RG122E10.2a; WUGSC:H_RG122E10.2b |
| Gene Symbols | RHBDD2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.