Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RHBDD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560571
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560571 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560571 Supplier Invitrogen™ Supplier No. PA560571
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HWEDSALPPP SLRPVQPTWE GSSEAGLDWA GASFSPGTPM WAALDEQMLQ EGIQASLLDG PAQEPQSAPW LSKSSVSSL Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 62%.

The Rhomboid family of proteins is made up of several widely conserved polytopic membrane serine proteases that play roles in growth and development. RHBDD3 was initially identified as a novel pituitary tumor apoptosis gene (PTAG) whose expression was reduced in certain pituitary adenomas. Overexpression of RHBDD3 in AtT20 cells showed an increase in apoptotic activity and caspase activation in response to bromocriptine, and apoptosis-inducing dopamine D2 analog, suggesting that reactivation of RHBDD3 in tumors may be a therapeutically useful tool. Later experiments also showed that RHBDD3 expression is also reduced in several primary colorectal tumors, indicating that loss of RHBDD3 contributes to a blunted apoptotic response and probably predisposes cells towards malignant transformation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RHBDD3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RHBDD3
Gene Accession No. Q9Y3P4
Gene Alias C22orf3; CTA-984G1.4; HS984G1A; pituitary tumor apoptosis; pituitary-derived proapoptotic; PTAG; RGD1311827; Rhbdd3; rhomboid domain containing 3; rhomboid domain-containing protein 3
Gene Symbols RHBDD3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RHBDD3. Recombinant protein control fragment (Product #RP-99855).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25807
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.