Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RHBDF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110292
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110292 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110292 Supplier Invitrogen™ Supplier No. PA5110292
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HRLPRRRKRE SVAKMSFRAA AALMKGRSVR DGTFRRAQRR SFTPASFLEE DTTDFPDELD TSFFAREGIL HEELSTYPDE VFESPSEAAL KDWEKAPEQA DLTGGALDRS ELERSHLMLP LERGWRKQKE GAAAPQPKVR LRQEVVSTAG PRRGQRIAVP VRKLFAREKR PYGLGMVGRL TNRTYRKRID SFVKRQIEDM D.

RHBDF1 is a seven-transmembrane protein with a long N-terminal cytoplasmic extension that comprises half of the polypeptide sequence, and is found in the endoplasmic reticulum and Golgi, but not on the cell surface. RHBDF1 has a pivotal role in sustaining growth signals in epithelial cancer cells and thus may serve as a therapeutic target for treating epithelial cancers.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RHBDF1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.03 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene RHBDF1
Gene Accession No. Q499S9, Q6PIX5
Gene Alias C16orf8; Dist; Dist1; EGFR-RS; epidermal growth factor receptor related sequence; epidermal growth factor receptor, related sequence; epidermal growth factor receptor-related protein; FLJ2235; FLJ22357; gene -89; gene -90; gene-89; gene-90; hDist1; hypothetical protein LOC431775; inactive rhomboid protein 1; Irhom1; Kiaa4242; mKIAA4242; p100hRho; Rhbdf1; rhbdf1a; Rhomboid 5 homolog 1; rhomboid 5 homolog 1 (Drosophila); rhomboid 5 homolog 1a (Drosophila); rhomboid family 1; rhomboid family 1 (Drosophila); rhomboid family member 1; zgc:91984
Gene Symbols RHBDF1
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13650, 303008
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.