Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RHBDF1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA543410
Description
Peptide sequence: KDWEKAPEQA DLTGGALDRS ELERSHLMLP LERGWRKQKE GAAAPQPKVR Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 79%.
RHBDF1 is a seven-transmembrane protein with a long N-terminal cytoplasmic extension that comprises half of the polypeptide sequence, and is found in the endoplasmic reticulum and Golgi, but not on the cell surface. RHBDF1 has a pivotal role in sustaining growth signals in epithelial cancer cells and thus may serve as a therapeutic target for treating epithelial cancers.
Specifications
RHBDF1 | |
Polyclonal | |
Unconjugated | |
RHBDF1 | |
C16orf8; Dist; Dist1; EGFR-RS; epidermal growth factor receptor related sequence; epidermal growth factor receptor, related sequence; epidermal growth factor receptor-related protein; FLJ2235; FLJ22357; gene -89; gene -90; gene-89; gene-90; hDist1; hypothetical protein LOC431775; inactive rhomboid protein 1; Irhom1; Kiaa4242; mKIAA4242; p100hRho; Rhbdf1; rhbdf1a; Rhomboid 5 homolog 1; rhomboid 5 homolog 1 (Drosophila); rhomboid 5 homolog 1a (Drosophila); rhomboid family 1; rhomboid family 1 (Drosophila); rhomboid family member 1; zgc:91984 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
64285 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96CC6 | |
RHBDF1 | |
Synthetic peptide directed towards the N-terminal of human RHBDF1 (aa 290-339). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction