Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RHBDF2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5121461
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121461 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5121461 Supplier Invitrogen™ Supplier No. PA5121461
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody tested in Indirect ELISA.

Regulates ADAM17 protease, a sheddase of the epidermal growth factor (EGF) receptor ligands and TNF, thereby plays a role in sleep, cell survival, proliferation, migration and inflammation. Does not exhibit any protease activity on its own. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen RHBDF2
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 3.65 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene RHBDF2
Gene Accession No. Q6PJF5
Gene Alias Inactive rhomboid protein 2; IRHOM2; RHBDF2; RHBDL5; RHBDL6; rhomboid 5 homolog 2; Rhomboid family member 2; rhomboid veinlet-like protein 5; Rhomboid veinlet-like protein 6; TEC; TOC; TOCG
Gene Symbols RHBDF2
Host Species Rabbit
Immunogen Recombinant Human Inactive rhomboid protein 2 protein (1-200aa).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79651
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.