Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RHBDL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560519
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560519 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560519 Supplier Invitrogen™ Supplier No. PA560519
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YAVWKPQKQW ITLDTGILES PFIYSPEKRE EAWRFISY Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 32%.

RHBDL2 is a member of the rhomboid family of integral membrane proteins. This family contains proteins that are related to Drosophila rhomboid protein. Members of this family are found in both prokaryotes and eukaryotes and are thought to function as intramembrane serine proteases. RHBDL2 is thought to release soluble growth factors by proteolytic cleavage of certain membrane-bound substrates, including ephrin B2 and ephrin B3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RHBDL2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RHBDL2
Gene Accession No. Q9NX52
Gene Alias 9130416B15; CTF; NTF; RHBDL2; rhomboid (veinlet, Drosophila)-like 2; rhomboid like 2; rhomboid protease 2; rhomboid, veinlet-like 2; rhomboid, veinlet-like 2 (Drosophila); rhomboid-like protein 2; Rhomboid-related protein 2; Rhomboid-related protein 2, C-terminal fragment; Rhomboid-related protein 2, N-terminal fragment; RRP2
Gene Symbols RHBDL2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RHBDL2. Recombinant protein control fragment (Product #RP-100813).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54933
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.