Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RHBG Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5142606
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142606 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5142606 Supplier Invitrogen™ Supplier No. PA5142606
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:2,000. Preliminary analysis showed no signal in Western Blot.

RHBG encodes one of two non-erythroid members of the Rhesus (Rh) protein family. Non-erythroid Rh protein family members are mainly expressed in the kidney and belong to the methylammonium-ammonium permease/ammonia transporters superfamily. All Rh family proteins are predicted to be transmembrane proteins with 12 membrane spanning domains and intracytoplasmic N- and C-termini.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RHBG
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene RHBG
Gene Accession No. Q9H310
Gene Alias Ammonium transporter Rh type B; Rh family B glycoprotein; Rh family B glycoprotein (gene/pseudogene); rh family type B glycoprotein; Rh family, B glycoprotein; Rh type B glycoprotein; Rhbg; Rhesus blood group family type B glycoprotein; Rhesus blood group, B glycoprotein; Rhesus blood group-associated B glycoprotein; SLC42A2
Gene Symbols RHBG
Host Species Goat
Immunogen Peptide with sequence C-PQLEKSKHRQ.
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57127
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.