Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RHBG Polyclonal Antibody, Invitrogen™

Catalog No. PIPA543367
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543367 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543367 Supplier Thermo Scientific Supplier No. PA543367

Rabbit Polyclonal Antibody

Peptide sequence: LATHEAYGDG LESVFPLIAE GQRSATSQAM HQLFGLFVTL MFASVGGGLG Sequence homology: Cow: 100%; Dog: 79%; Guinea Pig: 100%; Horse: 86%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 93%

RHBG Polyclonal antibody specifically detects RHBG in Human samples. It is validated for Immunohistochemistry, Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RHBG
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RHBG
Gene Accession No. Q9H310
Gene Alias Ammonium transporter Rh type B; Rh family B glycoprotein; Rh family B glycoprotein (gene/pseudogene); rh family type B glycoprotein; Rh family, B glycoprotein; Rh type B glycoprotein; Rhbg; Rhesus blood group family type B glycoprotein; Rhesus blood group, B glycoprotein; Rhesus blood group-associated B glycoprotein; SLC42A2
Gene Symbols RHBG
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human RHBG
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57127
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.