Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RHCE Antibody (BRIC69), Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $499.50
Specifications
Antigen | RHCE |
---|---|
Clone | BRIC69 |
Concentration | 1.0 mg/mL |
Dilution | Flow Cytometry 1:10-1:1000, Immunoprecipitation 1:10-1:500 |
Applications | Flow Cytometry, Immunoprecipitation |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017751
![]() |
Novus Biologicals
NB100658870.025MG |
0.025 mg |
Each for $206.50
|
|
|||||
NB10065887
![]() |
Novus Biologicals
NB10065887 |
0.1 mg |
Each for $499.50
|
|
|||||
Description
RHCE Monoclonal specifically detects RHCE in Human samples. It is validated for Flow Cytometry, Immunoprecipitation.Specifications
RHCE | |
1.0 mg/mL | |
Flow Cytometry, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Immunology | |
P18577 | |
6006 | |
Human erythrocytes | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
BRIC69 | |
Flow Cytometry 1:10-1:1000, Immunoprecipitation 1:10-1:500 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
blood group Rh(CE) polypeptide, blood group RhCcEe antigen, CD240CE, CD240CE antigen, MGC103977, RH, Rh blood group antigen Evans, Rh blood group C antigen, Rh blood group, CcEe antigens, Rh polypeptide 1, Rh polypeptide I, Rh30A, Rh4, RHC, RHCE blood group variant Crawford antigen Rh43, RHE, Rhesus blood group CE protein, Rhesus blood group E antigen, Rhesus blood group Rhce antigen, Rhesus blood group, CcEe antigens, Rhesus C/E antigens, Rhesus system C and E polypeptides, RhIVb(J), RHIXB, RhPI, RhVI, RhVIII, silenced Rh blood group CcEe antigen | |
RHCE | |
IgG1 | |
Protein A or G purified | |
Recognizes human erythrocytes of normal Rh types but does not recognize Rh null erythrocytes. NB100-65887 reacts with 31kD and 35-52kD proteins which were designated CD240DCE at the 7th leucocyte typing workshop. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title