Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RHCE Antibody (BRIC69), PE/Cy5.5, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10065887PECY55
Description
RHCE Monoclonal specifically detects RHCE in Human samples. It is validated for Flow Cytometry.Specifications
| RHCE | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| PBS with 0.05% Sodium Azide | |
| RHCE | |
| Human erythrocytes | |
| 0.1 mL | |
| Primary | |
| Recognizes human erythrocytes of normal Rh types but does not recognize Rh null erythrocytes. NB100-65887 reacts with 31kD and 35-52kD proteins which were designated CD240DCE at the 7th leucocyte typing workshop. | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| BRIC69 | |
| Flow Cytometry | |
| blood group Rh(CE) polypeptide, blood group RhCcEe antigen, CD240CE, CD240CE antigen, MGC103977, RH, Rh blood group antigen Evans, Rh blood group C antigen, Rh blood group, CcEe antigens, Rh polypeptide 1, Rh polypeptide I, Rh30A, Rh4, RHC, RHCE blood group variant Crawford antigen Rh43, RHE, Rhesus blood group CE protein, Rhesus blood group E antigen, Rhesus blood group Rhce antigen, Rhesus blood group, CcEe antigens, Rhesus C/E antigens, Rhesus system C and E polypeptides, RhIVb(J), RHIXB, RhPI, RhVI, RhVIII, silenced Rh blood group CcEe antigen | |
| Mouse | |
| Protein A or G purified | |
| Immunology | |
| 6006.0 | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction