Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RHCE Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP286778
Description
RHCE Polyclonal specifically detects RHCE in Human samples. It is validated for Western Blot.Specifications
RHCE | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
blood group Rh(CE) polypeptide, blood group RhCcEe antigen, CD240CE, CD240CE antigen, MGC103977, RH, Rh blood group antigen Evans, Rh blood group C antigen, Rh blood group, CcEe antigens, Rh polypeptide 1, Rh polypeptide I, Rh30A, Rh4, RHC, RHCE blood group variant Crawford antigen Rh43, RHE, Rhesus blood group CE protein, Rhesus blood group E antigen, Rhesus blood group Rhce antigen, Rhesus blood group, CcEe antigens, Rhesus C/E antigens, Rhesus system C and E polypeptides, RhIVb(J), RHIXB, RhPI, RhVI, RhVIII, silenced Rh blood group CcEe antigen | |
The immunogen is a synthetic peptide directed towards the middle region of human RHCE | |
100 μL | |
Immunology | |
6006 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction